4-Amino-5-bromo-2-chloropyridine ≥95%
Supplier: Matrix Scientific
Synonyms:
5-bromo-2-chloropyridin-4-amine, 5-Bromo-2-chloro-4-pyridylamine, 5-Bromo-2-chloro-4-pyridinamine
047196-1G
047196-5G
101878-884EA
92.31
USD
101878-884
102551-366
4-Amino-5-bromo-2-chloropyridine ≥95%
4-Amino-5-bromo-2-chloropyridine
Hazard code(s): Toxic, Store in Freezer
Formula:
C₅H₄BrClN₂ MW: 207.46 g/mol Melting Pt: 127…128 °C |
MDL Number:
MFCD09037459 CAS Number: 857730-21-3 |
Specification Test Results
Melting point | 156-157°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...