5-Chloro-2-formylpyridine ≥95% for synthesis

Supplier: Matrix Scientific

Synonyms: 5-Chloropyridine-2-carbaldehyde, 5-Chloropicolinaldehyde, 5-Chloro-2-pyridinecarbaldehyde

046969-1G 046969-5G 046969-25G
101877-846EA 64.62 USD
101877-846 102551-354 102551-356
5-Chloro-2-formylpyridine ≥95% for synthesis
5-Chloro-2-formylpyridine

Hazard code(s): Irritant

Formula: C₆H₄ClNO
Melting Pt: 66…68 °C
MDL Number: MFCD08277271
CAS Number: 31181-89-2

Order Now

Specification Test Results

Melting point 66-68°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR