2-Amino-3-iodo-5-nitropyridine ≥95% for synthesis

Supplier: Matrix Scientific

Synonyms: 3-Iodo-5-nitropyridin-2-amine, 3-Iodo-5-nitro-2-pyridylamine

046760-500MG 046760-1G
101876-900EA 24.62 USD
101876-900 101876-902
2-Amino-3-iodo-5-nitropyridine ≥95% for synthesis
2-Amino-3-iodo-5-nitropyridine

Hazard code(s): Irritant

Formula: C₅H₄IN₃O₂
MW: 265.01
Melting Pt: 220 °C
MDL Number: MFCD07368863
CAS Number: 25391-56-4

Order Now

Specification Test Results

Melting point 220°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR