3-(Trifluoromethyl)benzoylacetonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Cyano-3'-(trifluoromethyl)acetophenone, 3-Oxo-3-[3-(trifluoromethyl)phenyl]propanenitrile, 3-(3-(Trifluoromethyl)phenyl)-3-oxopropanenitrile

046710-1G 046710-5G 046710-25G
101876-674EA 215.38 USD
101876-674 101876-676 101876-678
3-(Trifluoromethyl)benzoylacetonitrile ≥95%
3-(Trifluoromethyl)benzoylacetonitrile

Hazard code(s): Irritant

Formula: C₁₀H₆F₃NO
MW: 213.16 g/mol
Melting Pt: 64…66 °C
MDL Number: MFCD00052315
CAS Number: 27328-86-5

Order Now

Specification Test Results

Melting point 64-66°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR