2-Chloro-6-nitrobenzothiazole ≥96%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-6-nitrobenzo[d]thiazole, 2-Chloro-6-nitro-1,3-benzothiazole

046366-1G 046366-5G
101875-190EA 40 USD
101875-190 101875-192
2-Chloro-6-nitrobenzothiazole ≥96%
2-Chloro-6-nitrobenzothiazole

Hazard code(s): Irritant

Formula: C₇H₃ClN₂O₂S
MW: 214.63 g/mol
Melting Pt: 192…195 °C
MDL Number: MFCD00022852
CAS Number: 2407-11-6
EINECS: 219-302-0

Order Now

Specification Test Results

Melting point 192-195°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR