2-Bromo-6-fluorobenzaldehyde ≥95%

Supplier: Matrix Scientific

046365-5G 046365-25G
101875-184EA 27.69 USD
101875-184 101875-186
2-Bromo-6-fluorobenzaldehyde ≥95%
2-Bromo-6-fluorobenzaldehyde

Hazard code(s): Irritant, Store in Freezer

Formula: C₇H₄BrFO
MW: 203.01 g/mol
Melting Pt: 45…48 °C
MDL Number: MFCD03407341
CAS Number: 360575-28-6

Order Now

Specification Test Results

Melting point 45-48°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR