Methyl-2-chloro-6-methoxynicotinate ≥95% for synthesis

Supplier: Matrix Scientific

Synonyms: Methyl 2-chloro-6-methoxynicatinate, Methyl 2-chloro-6-methoxynicotinate

046234-500MG 046234-1G
101874-768EA 141.54 USD
101874-768 101874-770
Methyl-2-chloro-6-methoxynicotinate ≥95% for synthesis
Methyl-2-chloro-6-methoxynicotinate

Hazard code(s): Irritant

Formula: C₈H₈ClNO₃
Melting Pt: 67…69 °C
MDL Number: MFCD08275100
CAS Number: 95652-77-0

Order Now

Specification Test Results

Melting point 67-69°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR