3-Bromo-2-fluorobenzaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Bromo-2-fluorobenzenecarbaldehyde

046062-5G 046062-25G
101873-970EA 44.62 USD
101873-970 102833-138
3-Bromo-2-fluorobenzaldehyde ≥95%
3-Bromo-2-fluorobenzaldehyde

Hazard code(s): Irritant

Formula: C₇H₄BrFO
Melting Pt: 43…46 °C
MDL Number: MFCD09027089
CAS Number: 149947-15-9

Order Now

Specification Test Results

Melting point 43-46°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR