3-Chlorobenzoylacetonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 3'-Chloro-2-cyanoacetophenone, (3-Chlorobenzoyl)acetonitrile, 3-(3-Chlorophenyl)-3-oxopropanenitrile

046056-1G 046056-5G
101873-944EA 40 USD
101873-944 101873-946
3-Chlorobenzoylacetonitrile ≥95%
3-Chlorobenzoylacetonitrile

Hazard code(s): Irritant

Formula: C₉H₆ClNO
MW: 179.61 g/mol
Melting Pt: 82…84 °C
MDL Number: MFCD00067891
CAS Number: 21667-62-9

Order Now

Specification Test Results

Melting point 82-84°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR