6-Phenyl-3-formylpyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Phenyl-5-pyridinecarboxaldehyde, 6-Phenylnicotinaldehyde

046005-500MG 046005-1G 046005-5G
101873-692EA 307.69 USD
101873-692 101873-694 101873-696
6-Phenyl-3-formylpyridine ≥95%
6-Phenyl-3-formylpyridine

Hazard code(s): Irritant

Formula: C₁₂H₉NO
MW: 183.21
Melting Pt: 54…56 °C
MDL Number: MFCD03086130
CAS Number: 63056-20-2

Order Now

Specification Test Results

Melting point 54-56°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR