Methyl-2-chloro-5-iodonicotinate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 2-chloro-5-iodonicotinate, Methyl 2-chloro-5-iodopyridine-3-carboxylate
046002-500MG
046002-1G
046002-5G
101873-674EA
118.46
USD
101873-674
101873-676
101873-678
Methyl-2-chloro-5-iodonicotinate ≥95%
Methyl-2-chloro-5-iodonicotinate
Hazard code(s): Irritant
Formula:
C₇H₅ClINO₂ MW: 297.48 g/mol Melting Pt: 70…73 °C |
MDL Number:
MFCD06659607 CAS Number: 78686-83-6 |
Specification Test Results
Melting point | 70-73°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...