4-Bromo-2-methyl-6-nitroaniline ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-6-nitro-o-toluidine, 2-Amino-5-bromo-3-nitrotoluene

045963-5G 045963-25G
101873-488EA 36.92 USD
101873-488 102551-340
4-Bromo-2-methyl-6-nitroaniline ≥95%
4-Bromo-2-methyl-6-nitroaniline

Hazard code(s): Irritant

Formula: C₇H₇BrN₂O₂
MW: 231.05 g/mol
Melting Pt: 145 °C
MDL Number: MFCD00052919
CAS Number: 77811-44-0

Order Now

Specification Test Results

Melting point 145°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR