2-Bromo-6-methyl-4-nitroaniline ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-3-bromo-5-nitrotoluene, 6-Bromo-4-nitro-o-toluidine

045962-500MG 045962-1G 045962-5G
101873-478EA 181.54 USD
101873-478 101873-480 101873-482
2-Bromo-6-methyl-4-nitroaniline ≥95%
2-Bromo-6-methyl-4-nitroaniline

Hazard code(s): Irritant

Formula: C₇H₇BrN₂O₂
MW: 231.05 g/mol
Melting Pt: 170…172 °C
MDL Number: MFCD00053089
CAS Number: 102170-56-9

Order Now

Specification Test Results

Melting point 170-172°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR