Methyl-2-aminonicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-aminonicotinate

045903-5G 045903-25G
101873-182EA 36.92 USD
101873-182 101873-184
Methyl-2-aminonicotinate ≥95%
Methyl-2-aminonicotinate

Hazard code(s): Irritant

Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Melting Pt: 83…85 °C
MDL Number: MFCD00128872
CAS Number: 14667-47-1

Order Now

Specification Test Results

Melting point 83-85°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR