4'-Methoxybiphenyl-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4'-Methoxy-[1,1'-biphenyl]-4-carboxylic acid, 4'-Methoxy[1,1'-biphenyl]-4-carboxylic acid

045780-500MG 045780-1G 045780-5G
101872-554EA 53.85 USD
101872-554 101872-556 101872-558
4'-Methoxybiphenyl-4-carboxylic acid ≥95%
4'-Methoxybiphenyl-4-carboxylic acid

Hazard code(s): Irritant

Formula: C₁₄H₁₂O₃
MW: 228.25 g/mol
Melting Pt: 253…256 °C
MDL Number: MFCD00185792
CAS Number: 725-14-4

Order Now

Specification Test Results

Melting point 253-256°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR