3-Amino-4-ethylcaproic acid ≥95% for synthesis

Supplier: Matrix Scientific

045692-500MG 045692-1G 045692-5G
101872-132EA 278.46 USD
101872-132 101872-134 101872-136
3-Amino-4-ethylcaproic acid ≥95% for synthesis
3-Amino-4-ethylcaproic acid

Hazard code(s): Irritant

Formula: C₈H₁₇NO₂
Melting Pt: 204…206 °C
MDL Number: MFCD01863280
CAS Number: 204191-42-4

Order Now

Specification Test Results

Melting point 204-206°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR