7-Nitro-1H-indazole ≥95%

Supplier: Matrix Scientific

Synonyms: 7-Nitroindazole, 7-NINA, 7-Nitro-1H-indazole

045648-500MG 045648-1G 045648-5G
101871-920EA 181.54 USD
101871-920 101871-922 101871-924
7-Nitro-1H-indazole ≥95%
7-Nitro-1H-indazole

Hazard code(s): Irritant

Formula: C₇H₅N₃O₂
MW: 163.14 g/mol
Melting Pt: 177…179 °C
Density: 1.56 g/cm³ (293 °C)
MDL Number: MFCD00022789
CAS Number: 2942-42-9
EINECS: 220-934-4

Order Now

Specification Test Results

Melting point 177-179°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR