3-Chloro-2-hydrazinopyridine ≥95%

Supplier: Matrix Scientific

045542-1G 045542-5G
101871-384EA 15.38 USD
101871-384 101871-386
3-Chloro-2-hydrazinopyridine ≥95%
3-Chloro-2-hydrazinopyridine

Hazard code(s): Irritant

Formula: C₅H₆ClN₃
Melting Pt: 165…167 °C
MDL Number: MFCD01936479
CAS Number: 22841-92-5

Order Now

Specification Test Results

Melting point 165-167°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR