2-Bromo-4-methyl-6-nitroaniline ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Bromo-6-nitro-p-toluidine, 4-Amino-3-bromo-5-nitrotoluene
045534-500MG
045534-1G
045534-5G
101871-344EA
278.46
USD
101871-344
101871-346
101871-348
2-Bromo-4-methyl-6-nitroaniline ≥95%
2-Bromo-4-methyl-6-nitroaniline
Hazard code(s): Irritant
Formula:
C₇H₇BrN₂O₂ Melting Pt: 63…65 °C |
MDL Number:
MFCD00209452 CAS Number: 827-24-7 |
Specification Test Results
Melting point | 63-65°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...