N-(2-Nitrobenzoyl)morpholine ≥95%

Supplier: Matrix Scientific

Synonyms: Morpholino(2-nitrophenyl)methanone

045483-500MG 045483-1G 045483-5G
101871-068EA 181.54 USD
101871-068 101871-070 101871-072
N-(2-Nitrobenzoyl)morpholine ≥95%
N-(2-Nitrobenzoyl)morpholine

Hazard code(s): Irritant

Formula: C₁₁H₁₂N₂O₄
Melting Pt: 117…119 °C
MDL Number: MFCD00586005
CAS Number: 26162-89-0

Order Now

Specification Test Results

Melting point 117-119°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR