1-Bromo-2,4-difluoro-5-nitrobenzene ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-2,4-difluoronitrobenzene

045474-5G 045474-25G 045474-100G
101871-014EA 44.62 USD
101871-014 101871-016 101871-018
1-Bromo-2,4-difluoro-5-nitrobenzene ≥95%
1-Bromo-2,4-difluoro-5-nitrobenzene

Hazard code(s): Irritant

Formula: C₆H₂BrF₂NO₂
MW: 237.99 g/mol
Boiling Pt: 85 °C (0.4mm)
MDL Number: MFCD00129166
CAS Number: 345-24-4

Order Now

Specification Test Results

Boiling point 85°C/0.4mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR