3-Furoyl chloride ≥95%

Supplier: Matrix Scientific

045463-500MG 045463-1G 045463-5G
101870-950EA 107.69 USD
101870-950 101870-952 101870-954
3-Furoyl chloride ≥95%
3-Furoyl chloride

Hazard code(s): Corrosive, Lachrymator, Moisture Sensitive, Stored under Argon

Formula: C₅H₃ClO₂
Boiling Pt: 62…64 °C (15mm)
MDL Number: MFCD02681970
CAS Number: 26214-65-3

Order Now

Specification Test Results

Boiling point 65°C/25mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR