N-(4-Nitrobenzoyl)morpholine ≥95%

Supplier: Matrix Scientific

Synonyms: Morpholino(4-nitrophenyl)methanone

045460-500MG 045460-1G 045460-5G
101870-938EA 181.54 USD
101870-938 101870-940 101870-942
N-(4-Nitrobenzoyl)morpholine ≥95%
N-(4-Nitrobenzoyl)morpholine

Hazard code(s): Irritant

Formula: C₁₁H₁₂N₂O₄
MW: 236.23 g/mol
Melting Pt: 100…103 °C
MDL Number: MFCD00451121
CAS Number: 5397-76-2

Order Now

Specification Test Results

Melting point 100-103°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR