2-Amino-5,6-dimethyl-4-hydroxypyrimidine ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Amino-5,6-dimethyl-4-pyrimidinol
045458-500MG
045458-1G
045458-5G
101870-926EA
278.46
USD
101870-926
101870-928
101870-930
2-Amino-5,6-dimethyl-4-hydroxypyrimidine ≥95%
2-Amino-5,6-dimethyl-4-hydroxypyrimidine
Hazard code(s): Irritant
Formula:
C₆H₉N₃O MW: 139.16 g/mol Melting Pt: 341 °C |
MDL Number:
MFCD00023258 CAS Number: 3977-23-9 EINECS: 223-611-6 |
Specification Test Results
Melting point | 341°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...