Methyl-5-bromo-6-chloronicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 5-bromo-6-chloronicotinate

045436-1G 045436-5G
101870-818EA 15.38 USD
101870-818 101870-820
Methyl-5-bromo-6-chloronicotinate ≥95%
Methyl-5-bromo-6-chloronicotinate

Hazard code(s): Irritant

Formula: C₇H₅BrClNO₂
MW: 250.48 g/mol
Boiling Pt: 279 °C
Melting Pt: 75…77 °C
Density: 1.684 g/cm³
MDL Number: MFCD03844855
CAS Number: 78686-77-8

Order Now

Specification Test Results

Melting point 75-77°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR