1-(2-Fluoro-4-nitrophenyl)piperazine ≥95%

Supplier: Matrix Scientific

045393-1G 045393-5G
101870-604EA 80 USD
101870-604 101870-606
1-(2-Fluoro-4-nitrophenyl)piperazine ≥95%
1-(2-Fluoro-4-nitrophenyl)piperazine

Hazard code(s): Irritant

Formula: C₁₀H₁₂FN₃O₂
MW: 225.22 g/mol
Melting Pt: 70…72.5 °C
MDL Number: MFCD01572463
CAS Number: 154590-33-7

Order Now

Specification Test Results

Melting point 70-72.5°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR