1-Methylindol-3-ylacetic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
1-Methyl-3-indoleacetic acid, 2-(1-Methyl-1H-indol-3-yl)acetic acid
045314-500MG
045314-1G
045314-5G
101870-256EA
181.54
USD
101870-256
101870-258
101870-260
1-Methylindol-3-ylacetic acid ≥95%
1-Methylindol-3-ylacetic acid
Hazard code(s): Irritant
Formula:
C₁₁H₁₁NO₂ MW: 189.21 g/mol Melting Pt: 133…135 °C |
MDL Number:
MFCD00130160 CAS Number: 1912-48-7 |
Specification Test Results
Melting point | 133-135°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...