2,3-Dichloroisonicotinic acid ≥95%
Supplier: Matrix Scientific
045193-250MG
045193-1G
101869-618EA
63.08
USD
101869-618
101869-620
2,3-Dichloroisonicotinic acid ≥95%
2,3-Dichloroisonicotinic acid
Hazard code(s): Irritant
Formula:
C₆H₃Cl₂NO₂ Melting Pt: 187 °C |
MDL Number:
MFCD01861977 CAS Number: 184416-84-0 |
Specification Test Results
Melting point | 187°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...