Methyl-2-chloro-5-nitrobenzoate ≥95% for synthesis

Supplier: Matrix Scientific

Synonyms: 2-Chloro-5-nitrobenzoic acid methyl ester, Methyl 2-chloro-5-nitrobenzenecarboxylate

045139-5G 045139-25G
101869-320EA 36.92 USD
101869-320 101869-322
Methyl-2-chloro-5-nitrobenzoate ≥95% for synthesis
Methyl-2-chloro-5-nitrobenzoate

Hazard code(s): Irritant

Formula: C₈H₆ClNO₄
Melting Pt: 70…72 °C
MDL Number: MFCD00184587
CAS Number: 6307-82-0

Order Now

Specification Test Results

Melting point 70-72°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR