1-(4-Amino-2-fluorophenyl)-4-methylpiperazine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Fluoro-4-(4-methylpiperazino)aniline, 3-fluoro-4-(4-methyl-1-piperazinyl)benzenamine, 3-Fluoro-4-(4-methylpiperazin-1-yl)aniline

045112-500MG 045112-1G 045112-5G
101869-200EA 24.62 USD
101869-200 101869-202 101869-204
1-(4-Amino-2-fluorophenyl)-4-methylpiperazine ≥95%
1-(4-Amino-2-fluorophenyl)-4-methylpiperazine

Hazard code(s): Irritant

Formula: C₁₁H₁₆FN₃
MW: 209.27 g/mol
Melting Pt: 87…89 °C
MDL Number: MFCD05663793
CAS Number: 221198-99-8

Order Now

Specification Test Results

Melting point 87-89°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR