(4-Methoxybenzyl)malononitrile ≥95%

Supplier: Matrix Scientific

045044-500MG 045044-1G 045044-5G
101868-874EA 375.38 USD
101868-874 101868-876 101868-878
(4-Methoxybenzyl)malononitrile ≥95%
(4-Methoxybenzyl)malononitrile

Hazard code(s): Irritant

Formula: C₁₁H₁₀N₂O
Melting Pt: 88…91 °C
MDL Number: MFCD00231623
CAS Number: 5553-92-4

Order Now

Specification Test Results

Melting point 88-91°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR