Acetophenone oxime ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl phenyl ketoxime, 1-Phenyl-1-ethanone oxime, Acetophenone Oxime

044998-500MG 044998-1G 044998-5G
101868-636EA 181.54 USD
101868-636 101868-638 101868-640
Acetophenone oxime ≥95%
Acetophenone oxime

Hazard code(s): Irritant

Formula: C₈H₉NO
MW: 135.17 g/mol
Melting Pt: 59…61 °C
MDL Number: MFCD00013931
CAS Number: 613-91-2
EINECS: 210-360-2

Order Now

Specification Test Results

Melting point 59-61°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR