Methyl-6-amino-5-bromonicotinate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 6-amino-5-bromonicotinate
044936-500MG
044936-1G
044936-5G
101868-310EA
375.38
USD
101868-310
101868-312
101868-314
Methyl-6-amino-5-bromonicotinate ≥95%
Methyl-6-amino-5-bromonicotinate
Hazard code(s): Irritant
Formula:
C₇H₇BrN₂O₂ MW: 231.05 g/mol Melting Pt: 166…168 °C |
MDL Number:
MFCD06659561 CAS Number: 180340-70-9 |
Specification Test Results
Melting point | 166-168°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...