Methyl-4,6-dichloronicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4,6-dichloronicotinate

044913-1G 044913-5G 044913-25G
101868-186EA 15.38 USD
101868-186 101868-188 101868-190
Methyl-4,6-dichloronicotinate ≥95%
Methyl-4,6-dichloronicotinate

Hazard code(s): Irritant

Formula: C₇H₅Cl₂NO₂
MW: 206.03 g/mol
Melting Pt: 41…43 °C
MDL Number: MFCD04125732
CAS Number: 65973-52-6

Order Now

Specification Test Results

Melting point 41-43°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR