2,6-Dichloro-3-nitrobenzonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 2,6-Dichloro-3-nitrobenzenecarbonitrile

044892-1G 044892-5G
101868-088EA 36.92 USD
101868-088 101868-090
2,6-Dichloro-3-nitrobenzonitrile ≥95%
2,6-Dichloro-3-nitrobenzonitrile

Hazard code(s): Irritant

Formula: C₇H₂Cl₂N₂O₂
Melting Pt: 106…109 °C
MDL Number: MFCD00051513
CAS Number: 5866-98-8
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 106-109°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR