Nicotinaldehyde oxime ≥95% for synthesis

Supplier: Matrix Scientific
Warning

Synonyms: 3-Pyridinealdoxime, Pyridine-3-carboxaldoxime, Pyridine-3-aldoxime

044844-500MG 044844-1G 044844-5G
101867-850EA 181.54 USD
101867-850 101867-852 101867-854
Nicotinaldehyde oxime ≥95% for synthesis
Nicotinaldehyde oxime
Formula: C₆H₆N₂O
MW: 122.13
MDL Number: MFCD00006409
CAS Number: 1193-92-6
EINECS: 214-782-8

Order Now

Specification Test Results

Melting point 148-151°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR