Methyl-6-hydroxynicotinate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 6-oxo-1,6-dihydro-3-pyridinecarboxylate, Methyl 6-hydroxynicotinate
044752-1G
044752-5G
101867-436EA
138.46
USD
101867-436
101867-438
Methyl-6-hydroxynicotinate ≥95%
Methyl-6-hydroxynicotinate
Hazard code(s): Irritant
Formula:
C₇H₇NO₃ MW: 153.14 g/mol Melting Pt: 166…170 °C |
MDL Number:
MFCD04113616 CAS Number: 66171-50-4 |
Specification Test Results
Melting point | 166-170°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...