4-Bromobenzoylacetonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(4-Bromophenyl)-3-oxopropanenitrile

044680-500MG 044680-1G 044680-5G
101867-070EA 15.38 USD
101867-070 101867-072 101867-074
4-Bromobenzoylacetonitrile ≥95%
4-Bromobenzoylacetonitrile

Hazard code(s): Irritant

Formula: C₉H₆BrNO
MW: 224.06 g/mol
Melting Pt: 161…163 °C
MDL Number: MFCD00996063
CAS Number: 4592-94-3

Order Now

Specification Test Results

Melting point 161-163°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR