2-Iodo-p-toluidine ≥95%

Supplier: Matrix Scientific

044675-5G 044675-25G
101867-044EA 76.92 USD
101867-044 102551-332
2-Iodo-p-toluidine ≥95%
2-Iodo-p-toluidine

Hazard code(s): Irritant

Formula: C₇H₈IN
Boiling Pt: 279 °C (1013 hPa)
Melting Pt: 39…41 °C
MDL Number: MFCD03407391
CAS Number: 29289-13-2
UN: 2811
ADR: 6.1,I

Order Now

Specification Test Results

Melting point 39-41°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR