4-Chloro-2-(trifluoromethoxy)aniline ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Chloro-2-(trifluoromethoxy)phenylamine

044671-1G 044671-5G
101867-018EA 98.46 USD
101867-018 101867-020
4-Chloro-2-(trifluoromethoxy)aniline ≥95%
4-Chloro-2-(trifluoromethoxy)aniline

Hazard code(s): Irritant

Formula: C₇H₅ClF₃NO
MW: 211.57 g/mol
MDL Number: MFCD00153102
CAS Number: 175205-77-3
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR