2-Hydroxy-4'-methoxyacetophenone ≥97%

Supplier: Matrix Scientific

044639-500MG 044639-1G 044639-5G
101866-846EA 275.38 USD
101866-846 101866-848 101866-850
2-Hydroxy-4'-methoxyacetophenone ≥97%
2-Hydroxy-4'-methoxyacetophenone

Hazard code(s): Irritant

Formula: C₉H₁₀O₃
Melting Pt: 100…103 °C
MDL Number: MFCD00004668
CAS Number: 4136-21-4
EINECS: 223-955-7

Order Now

Specification Test Results

Melting point 100-103°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR