2-Amino-3-bromo-5-chloropyridine ≥95%

Supplier: Matrix Scientific
Warning

Synonyms: 3-Bromo-5-chloro-2-pyridinamine, 3-Bromo-5-chloro-2-pyridylamine, 3-Bromo-5-chloropyridin-2-amine

044600-1G 044600-5G
101866-656EA 29.23 USD
101866-656 101866-658
2-Amino-3-bromo-5-chloropyridine ≥95%
2-Amino-3-bromo-5-chloropyridine

Hazard code(s): Irritant

Formula: C₅H₄BrClN₂
MW: 207.46 g/mol
Melting Pt: 82…83 °C
MDL Number: MFCD00955627
CAS Number: 26163-03-1

Order Now

Specification Test Results

Melting point 82-83°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR