5-Amino-1-methyl-1H-pyrazole ≥95%
Supplier: Matrix Scientific
Synonyms:
1-Methyl-1H-pyrazol-5-amine, 5-Amino-1-methylpyrazole
044579-1G
044579-5G
101866-552EA
15.38
USD
101866-552
101866-554
5-Amino-1-methyl-1H-pyrazole ≥95%
5-Amino-1-methyl-1H-pyrazole
Hazard code(s): Irritant
Formula:
C₄H₇N₃ MW: 97.12 g/mol Melting Pt: 72…74 °C |
MDL Number:
MFCD00068156 CAS Number: 1192-21-8 |
Specification Test Results
Melting point | 72-74°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...