4-Amino-1H-indazole ≥95%
Supplier: Matrix Scientific
044540-1G
044540-5G
101866-386EA
41.54
USD
101866-386
101866-388
4-Amino-1H-indazole ≥95%
4-Amino-1H-indazole
Hazard code(s): Irritant
Formula:
C₇H₇N₃ Melting Pt: 148…150 °C |
MDL Number:
MFCD03305710 CAS Number: 41748-71-4 |
Specification Test Results
Melting point | 148-150°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...