4-Nitro-1H-indazole ≥95%
Supplier: Matrix Scientific
044531-1G
044531-5G
101866-354EA
15.38
USD
101866-354
101866-356
4-Nitro-1H-indazole ≥95%
4-Nitro-1H-indazole
Hazard code(s): Irritant
Formula:
C₇H₅N₃O₂ Melting Pt: 199…203 °C |
MDL Number:
MFCD00022784 CAS Number: 2942-40-7 |
Specification Test Results
Melting point | 199-203°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...