2-Hydroxy-3'-nitroacetophenone ≥95% for synthesis

Supplier: Matrix Scientific

Synonyms: α-Hydroxy-3'-nitroacetophenone, 2-Hydroxy-1-(3-nitrophenyl)-1-ethanone

044325-500MG 044325-1G 044325-5G
101865-298EA 278.46 USD
101865-298 101865-300 101865-302
2-Hydroxy-3'-nitroacetophenone ≥95% for synthesis
2-Hydroxy-3'-nitroacetophenone

Hazard code(s): Irritant

Formula: C₈H₇NO₄
MW: 181.15
Melting Pt: 88…90 °C
MDL Number: MFCD04973723
CAS Number: 72802-41-6

Order Now

Specification Test Results

Melting point 88-90°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR