4'-Nitrobiphenyl-4-carboxylic acid ≥95%
Supplier: Matrix Scientific
044317-500MG
044317-1G
044317-5G
101865-258EA
272.31
USD
101865-258
101865-260
101865-262
4'-Nitrobiphenyl-4-carboxylic acid ≥95%
4'-Nitrobiphenyl-4-carboxylic acid
Hazard code(s): Irritant
Formula:
C₁₃H₉NO₄ MW: 243.22 g/mol Melting Pt: 350 °C |
MDL Number:
MFCD00043911 CAS Number: 92-89-7 EINECS: 202-201-0 |
Specification Test Results
Melting point | 350°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...