Methyl-5,6-dichloronicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 5,6-dichloronicotinate

044285-500MG 044285-1G 044285-5G
101865-100EA 18.46 USD
101865-100 101865-102 101865-104
Methyl-5,6-dichloronicotinate ≥95%
Methyl-5,6-dichloronicotinate

Hazard code(s): Irritant

Formula: C₇H₅Cl₂NO₂
MW: 206.03 g/mol
Melting Pt: 63…65 °C
MDL Number: MFCD00125367
CAS Number: 56055-54-0

Order Now

Specification Test Results

Melting point 63-65°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR