(2-Nitro-4-(trifluoromethyl)phenyl)acetic acid ≥95% for synthesis
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
2-[2-Nitro-4-(trifluoromethyl)phenyl]acetic acid
044276-5G
044276-25G
101865-048EA
75.38
USD
101865-048
101865-050
(2-Nitro-4-(trifluoromethyl)phenyl)acetic acid ≥95% for synthesis
(2-Nitro-4-(trifluoromethyl)phenyl)acetic acid
Hazard code(s): Irritant
Formula:
C₉H₆F₃NO₄ Melting Pt: 146…148 °C |
MDL Number:
MFCD00091534 CAS Number: 1735-91-7 EINECS: 217-081-5 |
Specification Test Results
Melting point | 146-148°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...