7-Amino-1H-indazole ≥95%

Supplier: Matrix Scientific

044189-500MG 044189-1G 044189-5G
101864-620EA 38.46 USD
101864-620 101864-622 101864-624
7-Amino-1H-indazole ≥95%
7-Amino-1H-indazole

Hazard code(s): Irritant

Formula: C₇H₇N₃
MW: 133.15 g/mol
Melting Pt: 156…159 °C
MDL Number: MFCD00022790
CAS Number: 21443-96-9
EINECS: 244-391-8

Order Now

Specification Test Results

Melting point 156-159°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR